Learn More
Axygen™ Automation Tips for Tecan™, Zymark™, Biotek™ and Dynamic Devices
Ensure consistent performance and quality. Axygen™ Automation Tips for Tecan™, Zymark™, Biotek™ and Dynamic Devices are constructed of clear polypropylene in 96-well racks. Available in nonsterile/sterile, non-filtered/filtered, wide-bore and bottom-labeled options with volumes ranging from 50 to 200μL.
Brand:  Axygen™ EVF-100-R-S
Code : 68
Additional Details : Weight : 9.50000kg
 View more versions of this product
Product Code. 12726689
  • £497.60 / Pack of 4800
Call for latest availability
+44 (0)1509 555 500
Add to basket
Description
Description
  • Comprehensive line of pipette tips for automated liquid handling systems
  • Manufactured to stringent specifications under strict process controls
  • Assembled via automation to ensure consistent performance and quality
  • Tips are certified RNase-/DNase-free and nonpyrogenic
  • Available in a range of volumes: 50, 100, 180 and 200μL
  • Compatible with the following:
    • Tecan™ Freedom Evo™, TeMo and Aquarius with MCA96 head
    • Zymark™/Caliper RapidPlate™, SciClone™ and Zephyr™ with HV96 head
    • Biotek™ Precision™ and Precision XS 8 or 12 head
    • Dynamic Devices Oasis LM 8/96-Channel-Standard Vol and 96-Channel-Low Vol head
Specifications
Specifications
Rack
Zymark™/Caliper™ RapidPlate™
Automation
Filtered
Clear
Round
ANSI
Non-pyrogenic, used for stringent specifications under strict process controls
96 Tips/Rack
Rack
100μL
Sterile
Light Gray
PP (Polypropylene)
DNase, Rnase Free
BioTek™ Precision™, BioTek™ Precision™ XS, Dynamic Devices Oasis LM, SciClone™, Tecan™ Aquarius, Tecan™ Freedom EVO®, Tecan™ TeMo, Zephyr™, Zymark™/Caliper™ RapidPlate™
Videos