missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Axygen™ Storage Microplates
For sample collection, in-vitro growth chambers and long term storage.
Brand: Axygen™ P-96-450R-C
Code : 68
Additional Details : Weight : 3.18000kg
Description
Axygen™ deep well and assay plates are used for sample collection, in vitro growth chambers and long-term storage. Made of polypropylene, they offer excellent chemical resistance and will withstand temperatures down to -80°C.Specifications
ANSI, SLAS 1-2004 | |
Without Lid | |
Autoclavable | |
Polypropylene | |
Non-sterile | |
Sample collection, in-vitro growth chambers and long term storage | |
50/Cs. | |
-112°F |
Standard | |
8 x 12 | |
Clear | |
96 | |
Round | |
4000 x g | |
10/Pk. | |
-80°C |