missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Axygen™ Storage Microplates
Brand: Axygen™ P-96-450V-W
Code : 68
Additional Details : Weight : 3.20000kg
Description
Choose from a variety of configurations:
- 48 deep-well rectangular plate with pyramid bottom
- 96 deep-well plate with square wells
- 384 deep-well diamond plate, with square wells
- 96-well assay plate with round bottom (SBS footprint) or V-bottom (SBS footprint)
- 24 deep-well rectangular plate
Specifications
ANSI, SLAS 1-2004 | |
8 x 12 | |
White | |
96 | |
Conical Bottom (V-well) | |
4000 x g | |
-80°C |
Standard | |
Autoclavable | |
Polypropylene | |
Non-sterile | |
Sample collection, in-vitro growth chambers and long term storage | |
-112°F |