Axygen™ Single Well Low Profile Reagent Reservoirs

Manufactured with virgin polypropylene for excellent chemical resistance

Overview
Brand: Axygen

Manufacturer Part Number: RES-SW96-LP-SI

UNSPSC: 24111800

Code: 68

Additional Details:
Additional Details: Weight: 2.70000kg



Product Code. 11390275

Quantity Price
1 £ 240.03 / Pack of 25
Call for latest availability
+44 (0)1509 555 500
Add to basket


Description and Specification

Specification

Disposable Yes
Product Type Reagent Reservoir
Material Polypropylene
Color Clear
Packaging Individually Wrapped; 25/Cs.
Capacity (Metric) 86mL
Includes 96-bottom troughs
Capacity 2.9 oz., 86mL
Sterile Yes
Certifications/Compliance ANSI/SLAS, RNase/DNase-free, non-pyrogenic