missing translation for 'onlineSavingsMsg'
Learn More

Axygen Cap looped, screw without ''O'' rings, polypropylene, fits 0.5mL, 1.5mL and 2.0mL tubes

Brand:  Axygen SC-LP-GR

Cap looped, screw without ''O'' rings, polypropylene, fits 0.5mL, 1.5mL and 2.0mL tubes

Code : 68

Additional Details : Weight : 2.70000kg

Product Code. 12749358

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.