missing translation for 'onlineSavingsMsg'
Learn More

Axygen Screw cap, grey, assorted sizes, fit 0.5mL, 1.5mL and 2.0mL tubes

Brand:  Axygen SC-GR

Screw cap, grey, assorted sizes, fit 0.5mL, 1.5mL and 2.0mL tubes

Code : 68

Additional Details : Weight : 2.30000kg

Product Code. 10329779

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.