missing translation for 'onlineSavingsMsg'
Learn More

Axygen™ Screw Caps with O-rings

Select looped or non-looped format

Brand:  Axygen SCO-G-S

UNSPSC : 41121710

Code : 68

Additional Details : Weight : 2.30000kg

 View more versions of this product

Product Code. 12739378

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Product Support team on 01509 555888 for assistance
View alternative products »


Specifications

Specifications

Polypropylene
Yes
Non-looped
Green
Both skirted and non-skirted variations of the 0.5mL, 1.5mL, and 2mL microcentrifuge tubes
O-ring