Learn More
Axygen™ Screw Caps with O-rings
Select looped or non-looped format
Brand:  Axygen™ SCO-B-S
Code : A0
Additional Details : Weight : 2.30000kg
 View more versions of this product
Product Code. 12518947
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Description
Description
Axygen microcentrifuge tubes feature a screw cap that provides a tight secure seal. Screw cap tubes are available in a variety of colors with a conical or self-standing bottom. Sterile options are also available.
  • Maximum G-force is 20,000 RCF.
  • RNase-/DNase-free
  • Nonpyrogenic
Tubes, with Cap, Sterile
All sterile screw cap microcentrifuge tubes are pre-assembled with non-looped caps with O-Rings.
Colored-cap versions are also available in sterile format.
Specifications
Specifications
O-ring
Blue
Sterile
Polypropylene
Non-looped
Both skirted and non-skirted variations of the 0.5 mL, 1.5 mL, and 2 mL microcentrifuge tubes