Learn More
Axygen™ Screw Cap Tubes without Caps: Self-Standing with Conical Bottoms
Self-standing tubes
Brand: Axygen™ ST-200-SS-S
Code : 68
Additional Details : Weight : 5.90000kg
Description
Serve a broad range of laboratory needs including collection, processing, centrifugation, storage and packaging/shipping
Specifications
20,000 x g | |
2 mL | |
Polypropylene | |
Sterile | |
Conical |
RNase, DNase, pyrogen-free | |
Clear | |
Bag | |
Not Included |