missing translation for 'onlineSavingsMsg'
Learn More

Axygen™ Screw Cap Tubes without Caps: Self-Standing

Self-standing tubes

Brand:  Axygen ST-150-SS

Code : 68

Additional Details : Weight : 6.35000kg

 View more versions of this product

Product Code. 12729608

  • £290.81 / Pack of 4000


Call for latest availability
+44 (0)1509 555 500
Add to basket

Description

Description

Serve a broad range of laboratory needs including collection, processing, centrifugation, storage and packaging/shipping

  • Tube volumes from 0.5 to 2.0mL
  • Choose clear or amber

Specifications

Specifications

RNase, DNase, pyrogen-free
1.5mL
Polypropylene
No
500 Tubes/Pk., 8 Packs/Cs.
Without
Clear
20000xG
No
Conical
Videos