Learn More
Axygen™ Screw Cap Tubes without Caps: Conical with Conical Bottoms
Conical tubes
Brand:  Axygen™ ST-150-C-S
Code : A0
Additional Details : Weight : 5.90000kg
 View more versions of this product
Product Code. 11324225
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Description
Description
Serve a broad range of laboratory needs including collection, processing, centrifugation, storage and packaging/shipping
Specifications
Specifications
RNase, DNase, pyrogen-free
Clear
Bag
Not Included
1.5 mL
Polypropylene
Sterile
Conical