Learn More
Axygen™ Qiagen™ Robotic Tips
Compatible with Qiagen liquid handling systems and Qiagen BioRobot Series
Brand:  Axygen QT-1100-CBK-R-S
Code : 68
Additional Details : Weight : 10.10000kg
 View more versions of this product
Product Code. 12555467
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Product Support team on 01509 555888 for assistance
View alternative products »
Specifications
Specifications
Qiagen™ BioRobot™
Automation
Filtered
Black
PP (Polypropylene)
DNase, Rnase Free
Qiagen™ BioRobot™
Rack
1100μL
Sterile
Round
Non-ANSI
Non-pyrogenic, liquid level sensing, used for stringent specifications under strict process controls
Videos