Axygen™ Polypropylene Universal Pipette Tip


Overview
Brand: Axygen

Manufacturer Part Number: TF-100-L

X10000 FILTER TIP 100µL, MAXYMUM RECEOVERY(Packs 10000Pcs)

UNSPSC: 41121601

Code: NEW

Additional Details:
Additional Details: Weight: 4.08233kg



Product Code. 13429808

 N/A
N/A

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Product Support team on 01509 555888 for assistance


  • Description and Specification
  • Alternative Products
Description and Specification

Specification

Quantity 10000Pack
For Use With (Equipment) For AxyPet-16 pipettes
Product Type Universal Pipette Tip
Material Polypropylene
Certifications/Compliance Certified to be RNase-/DNase-free
Volume (Metric) 1 to 100μL
Sterility Sterile
Autoclavable Yes
Format 100-Format
Packaging Bulk
Color Clear
Alternative Products