Learn More
Axygen™ Aerosol Barrier Micropipette Tips
Axygen™ Aerosol Barrier Micropipette Tips allow free passage of air while blocking aerosol contamination.
Brand:  Axygen™ TF-150
Code : 68
Additional Details : Weight : 3.97000kg
 View more versions of this product
Product Code. 12581348
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Description
Description
  • Technologically advanced filter material
  • Filter membrane contains no leachable additives
  • Available with Maxymum Recovery™ surface for improved accuracy and maximum sample recovery
  • Available in bulk, or racked (sterile)
  • Available in a variety of material contact tip options
Specifications
Specifications
100 to 1000 μL
Non-sterile
Autoclavable
Bulk
Bulk = 1000 tips/pack
Universal Fit
Filtered
Clear
DNase, RNase Free
Universal
Aerosol Barrier Micropipette Tips
Polypropylene