Learn More
Axygen™ Polypropylene Non-sterile Automation Tip
Brand: Axygen EV-50-R-LAB
Code : NEW
Additional Details : Weight : 10.88621kg
Specifications
50 μL | |
Non-sterile | |
Rack | |
Tecan Freedom EVO, Tecan TeMo, Tecan Aquarius MCA96 head, Zymark/Caliper RapidPlate, SciClone, Zephyr HV96, BioTek Precision, BioTek Precision XS 8, BioTek Precision XS 12, Dynamic Devices Oasis LM 8 96-Channel | |
Polypropylene |
Non-Filtered | |
Clear | |
Automation, Round | |
Tip |