Learn More
Axygen™ Polypropylene Full Skirt 384 Well Rigi Plate PCR Microplates
Brand: Axygen PCR-384-RGD-W
Code : 68
Additional Details : Weight : 3.20000kg
Specifications
Yes | |
16 x 24 | |
Polypropylene | |
40 μL | |
50 μL | |
Yes | |
100/Cs. |
Standard | |
White | |
384 | |
Non-sterile | |
Flat Bottom (F-well) | |
Provide extra rigidity in thermal cycling applications | |
Yes |