Axygen™ Automation Tips for PerkinElmer™, 96-Well
Ideal for Janus™ MiniTrak, PlateTrak™ and Evolution P3. Axygen™ Automation Tips for PerkinElmer™, 96-Well are constructed of clear polypropylene. Available in sterile/non-sterile, filtered/non-filtered and wide bore options, from 20 to 200µL.
Overview
Brand: Axygen™
Manufacturer Part Number: PKF-180-R-S
UNSPSC: 41121608
Code: 68
Additional Details:
Additional Details: Weight: 10.00000kg
Product Code. 12766709
Quantity Price
1 £ 480.27 / Pack of 4800
Call for latest availability
+44 (0)1509 555 500
Add to basket
Description and Specification
Specification
For Use With (Equipment) PerkinElmer™ Packard™ Janus™/Evolution P3™/MiniTrak™/PlateTrak™ automated liquid handling systems and MDT-96 P235, MDT-96 P50 and MDT-384 30μL head
Sterility Sterile
Product Type Filter, Racked, Presterilized
Material Polypropylene
Quantity 96 Tips/Rack
Volume (Metric) 180μL
Packaging Rack
Color Clear
For Use With (Application) Non-pyrogenic, used for stringent specifications under strict process controls, nucleic acid, nuclease and endotoxin-safe for complete protection against contamination
Rack Color Green
Shape Round
Certifications/Compliance RNase, DNase-free, ANSI
Assembled via automation under strict process controls to ensure consistent performance and quality
  • Robotic tips use no mold-releasing agents
  • Nucleic acid, nuclease and endotoxin-safe for complete protection against contamination
  • Tips are certified RNase-/DNase-free and nonpyrogenic
  • Compatible with PerkinElmer™ MDT-96 P235, MDT-96 P50 and MDT-384 30µL head for:
    • Janus™
    • Evolution P3
    • Minitrak
    • PlateTrak™
Multiple options
  • Presterilized and non-sterile tips
  • With or without aerosol-resistant polyethylene filters
  • Wide bore series for working with viscous materials, genomic DNA or delicate cell cultures
  • Available in a range of volumes: 20, 30, 50, 180 and 200µL