Learn More
Axygen™ Automation Tips for PerkinElmer™, 96-Well
Ideal for Janus™ MiniTrak, PlateTrak™ and Evolution P3. Axygen™ Automation Tips for PerkinElmer™, 96-Well are constructed of clear polypropylene. Available in sterile/non-sterile, filtered/non-filtered and wide bore options, from 20 to 200µL.
Brand:  Axygen™ PKF-180-R-S
Code : A0
Additional Details : Weight : 10.00000kg
 View more versions of this product
Product Code. 11370235
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Product Support team on 01509 555888 for assistance
Description
Description
Assembled via automation under strict process controls to ensure consistent performance and quality
  • Robotic tips use no mold-releasing agents
  • Nucleic acid, nuclease and endotoxin-safe for complete protection against contamination
  • Tips are certified RNase-/DNase-free and nonpyrogenic
  • Compatible with PerkinElmer™ MDT-96 P235, MDT-96 P50 and MDT-384 30µL head for:
    • Janus™
    • Evolution P3
    • Minitrak
    • PlateTrak™
Multiple options
  • Presterilized and non-sterile tips
  • With or without aerosol-resistant polyethylene filters
  • Wide bore series for working with viscous materials, genomic DNA or delicate cell cultures
  • Available in a range of volumes: 20, 30, 50, 180 and 200µL
Specifications
Specifications
Rack
PerkinElmer™ Janus™þPerkinElmer™ Evolution P3™þPerkinElmer™ MiniTrak™þPerkinElmer™ PlateTrak™þPerkinElmer™ MDT-96 P235þPerkinElmer™ MDT-96 P50þPerkinElmer™ MDT-384 30μL
180μL
Sterile
Green
PP (Polypropylene)
DNase, Rnase Free
PerkinElmer™ Janus™, PerkinElmer™ Evolution P3™, PerkinElmer™ MiniTrak™, PerkinElmer™ PlateTrak™, PerkinElmer™ MDT-96 P235, PerkinElmer™ MDT-96 P50, PerkinElmer™ MDT-384 30μL
96 Tips/Rack
Rack
Filtered
Clear
Round
ANSI
Non-pyrogenic, used for stringent specifications under strict process controls, nucleic acid, nuclease and endotoxin-safe for complete protection against contamination
Videos