Learn More
Axygen™ PCR Tubes with 0.5 mL Flat Cap
Thin and consistent wall thickness for precise thermal transfer
Brand:  Axygen™ PCR-05-L-C
Code : 68
Additional Details : Weight : 5.10000kg
 View more versions of this product
Product Code. 12766869
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Description
Description
Quantitative PCR compatible tubes for a wide range of Real-Time PCR instruments.
  • Tubes with caps sealed full-length; insertion cap ensures tight fit, eliminates sample evaporation
  • Choose from a wide variety of colors
  • Certified RNase-/DNase-free
  • Nonpyrogenic
  • Nonsterile
  • Manufactured from polypropylene
Specifications
Specifications
RNase, DNase, pyrogen-free
0.5 mL
Polypropylene
Conical
Polypropylene
Tube
Clear
1000/Pk., 10Pk./Cs.
Standard 96-well blocks of real-time PCR instruments and thermal transfer applications
Non-sterile
Flat
Maxymum Recovery PCR Tube