missing translation for 'onlineSavingsMsg'
Learn More

Axygen™ Oasis™ Robotic Tips

For use with Oasis automated liquid handling systems

Brand:  Axygen™ OT-200-R

Code : A0

Additional Details : Weight : 9.50000kg

 View more versions of this product

Product Code. 12599207

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.


Description

Description

  • Tip format: 96 tip
  • Tip volume range: 50 μL to 200 μL
  • Tip material: Clear polypropylene
  • RNase-/DNase-/human gDNA-free
  • Nonpyrogenic
  • Available options:
    - With or without aerosol-resistant filters
    - Nonsterile or sterile
  • Compatible heads: 8- or 96-channel standard volume
  • Rack format: ANSI/SLAS
  • Rack material: Polypropylene
Specifications

Specifications

200 μL
Non-sterile
Blue
Rack
SBS Rack
Dynamic Devices Oasis
Polyethylene
Non-Filtered
Clear
DNase, RNase Free
Automation, Round
50 Racks/Cs.
Oasis
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers