Learn More
Axygen™ Oasis™ Robotic Tips
For use with Oasis automated liquid handling systems
Brand:  Axygen OT-200-R
UNSPSC : 41121608
Code : 68
Additional Details : Weight : 9.50000kg
 View more versions of this product
Product Code. 10338029
  • £436.68 / Pack of 4800
Call for latest availability
+44 (0)1509 555 500
Add to basket
Specifications Description & Specifications
Specifications
Nonsterile
96 Tips/Rack
Dynamic devices Oasis™ automated liquid handling system
Clear
Rack
Polypropylene
Robotic Tip
200μL
Blue
RNase, DNase-free