Learn More
Axygen™ Multimek Robotic Tips: 200μL
Fits Multimek «Type II« or CCS-style head
Brand:  Axygen MT-200-R
Code : 68
Additional Details : Weight : 10.89000kg
 View more versions of this product
Product Code. 12772876
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Product Support team on 01509 555888 for assistance
View alternative products »
Description
Description
Robotic tip's unique diamond-polished molding technology uses no mold-releasing agents. Minimizes wastage of precious samples and reagents while handling. Nucleic acid, nuclease and endotoxin-safe for complete protection against contamination.
  • Tip volume: 200μL
  • Tip color: Clear
  • Rack color: Red and blue
Order Info
Racked: 96 tips/rack, 10 racks/pack, 5 packs/case
Specifications
Specifications
Robotic Tip
Nonsterile
Blue
Racked
Rack
Non-ANSI
Beckman™ Coulter™ MultiMek™ automated system
200μL
Clear
Yes
Polypropylene
96 Tips/Rack
Unique diamond polished molding technology use no mold-releasing agents, minimizes wastage of precious samples and reagents while handling, nucleic acid, nuclease and endotoxin-safe for complete protection against contamination
Videos