Learn More
Axygen™ GentleFit™ Dual-Shot Filter Pipet Tips
Soft, leakproof seal provides a universal fit
Brand: Axygen TLF-200-R-S
Code : 66
Additional Details : Weight : 8.60000kg
Description
These filter tips are manufactured from two different resins. The thermoplastic elastomer (TPE) top is easy to seal, provides a soft eject, and is colored-coded by volume. The rigid, polypropylene body ensures straightness, affords glass-like clarity and is non-reactive and graduated.
- Versatile; dual-shot technology standardizes most research-grade pipetters
- Available in three sizes: 10, 200 and 1000μL
- Reduced sealing force
- Minimal reciprocal ejection force
- Economical; imposes less wear on pipetters
- Ergonomic design; soft tip requires less pressure to create an optimum seal
- Ideal for multichannel pipetters
- Color-coded top identifies volume
- Presterilized
- Certified to be free from RNase, DNase, and human DNA
Specifications
200 μL | |
Sterile | |
Polypropylene |
Filtered | |
Rack |