missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Axygen™ GEN3™ Multi-Barrier Pipette Tips
Offers the ultimate in protection against cross contamination using tri-filter technology
£505.29 - £619.00
Specifications
Filtered | Filtered |
---|---|
Sterility | Sterile |
Autoclavable | Not Autoclavable |
Clean Claims | DNase, RNase, DNA, PCR inhibitor, and Pyrogen Free |
Format | Rack |
Product Code | Brand | Volume (Metric) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Volume (Metric) | Price | Quantity & Availability | |||||
12812283
|
Axygen™
GEN3-1000-L-R-S |
100 to 1000 μL |
£570.00
Pack of 5000 |
Estimated Shipment: 09-07-2024
Log in to see stock. |
|||||
12892273
|
Axygen™
GEN3-20-L-R-S |
20 μL |
£565.00
Pack of 4800 |
Estimated Shipment: 02-07-2024
Log in to see stock. |
|||||
12822453
|
Axygen™
GEN3-1000-L-R-S |
100 to 1000 μL |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
12882273
|
Axygen™
GEN3-200-L-R-S |
1 to 200 μL |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
12802283
|
Axygen™
GEN3-10-L-R-S |
0.1 to 10 μL |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Description
The new Axygen™ brand GEN3 multi-barrier filter pipet tips offer the ultimate in protection against cross contamination using tri-filter technology. These tips will prevent liquid or aerosol from passing through and will lock on contact. GEN3 tips effectively separate liquid from the self-sealing barrier using an additional protective layer of conventional polyethylene filter material. In addition to the self-sealing properties, our GEN3 multi-barrier pipet tips feature our exclusive Maxymum Recovery™ technology which reduces the amount of reagent or sample adhering to the pipet tips. Offered in Racks, sterile, tips are nonpyrogenic and certified human gDNA and PCR inhibition free and DNase-/RNase-free.Certifications
GEN3 multi-barrier tips are certified human gDNA and PCR inhibition free. Certified DNase-/RNase-free and nonpyrogenic.
Specifications
Filtered | |
Not Autoclavable | |
Rack | |
Polyethylene |
Sterile | |
DNase, RNase, DNA, PCR inhibitor, and Pyrogen Free | |
GEN3 |