Learn More
Axygen™ GEN3™ Multi-Barrier Pipette Tips
Offers the ultimate in protection against cross contamination using tri-filter technology
Brand: Axygen™ GEN3-10-L-R-S
Code : A0
Additional Details : Weight : 8.40000kg
Description
The new Axygen™ brand GEN3 multi-barrier filter pipet tips offer the ultimate in protection against cross contamination using tri-filter technology. These tips will prevent liquid or aerosol from passing through and will lock on contact. GEN3 tips effectively separate liquid from the self-sealing barrier using an additional protective layer of conventional polyethylene filter material. In addition to the self-sealing properties, our GEN3 multi-barrier pipet tips feature our exclusive Maxymum Recovery™ technology which reduces the amount of reagent or sample adhering to the pipet tips. Offered in Racks, sterile, tips are nonpyrogenic and certified human gDNA and PCR inhibition free and DNase-/RNase-free.Certifications
GEN3 multi-barrier tips are certified human gDNA and PCR inhibition free. Certified DNase-/RNase-free and nonpyrogenic.
Specifications
0.1 to 10 μL | |
Sterile | |
DNase, RNase, DNA, PCR inhibitor, and Pyrogen Free | |
96 Tips/Rack, 10 Racks/Pk, 5 Packs/Cs | |
Polyethylene |
Filtered | |
Not Autoclavable | |
Rack | |
GEN3 |