Learn More
Axygen™ Gel Tips, Round
Universal fit for research grade pipetters
Brand: Axygen™ TGL-200RD-57-R-S
Code : A0
Additional Details : Weight : 0.45000kg
Description
Axygen™ Gel Loading Tips are designed to fit a wide variety of single and multi-channel pipettors, compatible with most popular brand of pipettors. These polypropylene tips are nonpyrogenic and certified to be RNase-/DNase-free. Available in either flat or round versions, with different outer diameters for different gel thickness.Specifications
0.5 to 200 μL | |
Clear | |
Gel Loading, Round | |
Yes | |
2, 5μL |
Sterile | |
DNase, RNase Free | |
200 Tips/Rack, 2 Racks/Cs | |
Polypropylene |