missing translation for 'onlineSavingsMsg'
Learn More

Axygen™ Gel Cutting Tips: 1.1 x 6.5mm

Universal fit for research grade pipettes.

Brand:  Axygen™ TGL-1165

Code : 68

Additional Details : Weight : 2.72000kg

 View more versions of this product

Product Code. 12575717

  • £477.00 / Pack of 2500


Estimated Shipment
18-06-2021
Add to basket

Description

Description

Axygen™ Gel Cutting Tips are designed to remove bands from agarose gel. These polypropylene tips are nonpyrogenic and RNase-/DNase-free.

 

  • Designed for removing bands from agarose gel
  • One handed operation
  • Push button gel release
  • Ejector button tip release
  • Avoid cross contamination
  • Tips fit standard 1000 µL pipettors
  • These polypropylene tips are nonpyrogenic and RNase-/DNase-free.
  • Nonpyrogenic
  • Nonsterile
Specifications

Specifications

Non-sterile
2, 5μL
1.1 x 6.5 mm
Bulk
250 Tips/Pk, 10 Packs/Cs
Clear
PP (Polypropylene)
DNase, RNase Free
Gel Loading
Videos
SDS
Documents