Learn More
Axygen™ Gel Cutting Tips: 1.1 x 4 mm
Universal fit for research grade pipettes.
Brand: Axygen™ TGL-1140
Code : 68
Additional Details : Weight : 2.27000kg
Description
Axygen™ Gel Cutting Tips are designed to remove bands from agarose gel. These polypropylene tips are nonpyrogenic and RNase-/DNase-free.
- Designed for removing bands from agarose gel
- One handed operation
- Push button gel release
- Ejector button tip release
- Avoid cross contamination
- Tips fit standard 1000 µL pipettors
- These polypropylene tips are nonpyrogenic and certified to be RNase-/DNase-free.
- Nonpyrogenic
- Nonsterile
Specifications
Non-sterile | |
2, 5μL | |
1.1 x 4 mm | |
Bulk | |
250 Tips/Rack, 10 Packs/Cs |
Clear | |
PP (Polypropylene) | |
DNase, RNase Free | |
Gel Loading |