Learn More
Axygen PCR microplate 50µL working volume 384 well polypropylene for ABI 3730, 3100 and MJ Thermocyclers
Brand:  Axygen PCR-384M2-V
PCR microplate 50µL working volume 384 well polypropylene for ABI 3730, 3100 and MJ Thermocyclers
Code : 68
Additional Details : Weight : 1.40000kg
Product Code. 12779428