Learn More
Axygen™ FDSS™ Robotic Tips
For Hamamatsu FDSS series systems
Brand:  Axygen FDSS-384-BK-R
Code : A0
Additional Details : Weight : 10.10000kg
 View more versions of this product
Product Code. 12585717
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Product Support team on 01509 555888 for assistance
Description
Description
  • For 384-Well plates
  • Tip color: Black
  • Capacity: 50μL
  • Packaging: 384 Tips/rack; 10 Racks/unit; 5 Units/Cs.
Specifications
Specifications
Robotic Tip
Black
Racked
Rack
DNase, RNase, pyrogen free
50μL
Yes
Polypropylene
384 Tips/Rack
Hamamatsu FDSS™ 6000 system
Videos