Axygen™ Eppendorf™ Compatible Pipet Tips
Compatible with Eppendorf and Biohit pipets
Brand: Axygen
Manufacturer Part Number: TE-1004-B-L-R-S
X3456 Pipettor tip Axygen MAXYMUM RECOVERY pre-sterilised, racked (KZ use only)
UNSPSC: 41121607
Code: 68
Additional Details:
Additional Details: Weight: 6.80000kg
Product Code. 12746619
Quantity Price
1 £228.34 / Pack of 3456
Call for latest availability
+44 (0)1509 555 500
Add to basket
Description and Specification
Volume (Metric) 1000μL
Sterility Sterile
Color Blue
Graduations 110 and 500μL
Tip Style Eppendorf-style; Maxymum Recovery; Presterilized
Packaging Rack
  • Available in sterile or nonsterile forms
  • e-Beam irradiated to prevent product degradation
  • Maxymum recovery tips allows complete sample retrieval and greater tip-to-tip precision
  • Graduation marks at 110μL and 500μL, allows visual confirmation of pipettor draw volume
  • Volume: 1000μL
  • Color: Blue