Axygen™ Biomek™ FX/NX Robotic Tips


Overview
Brand: Axygen

Manufacturer Part Number: FX-384-L-R-S

UNSPSC: 41121608

Code: 68

Additional Details:
Additional Details: Weight: 10.40000kg



Product Code. 12702866

 N/A
N/A

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Product Support team on 01509 555888 for assistance


Description and Specification

Specification

Product Type Robotic Tip
Volume (Metric) 0.5 to 30μL
Sterility Sterile
Color Clear
Rack Color Green
Tip Style Maxymum Recovery
Shape Round
Material Polypropylene
Packaging Rack
Quantity 384 Tips/Rack
Certifications/Compliance RNase, DNase-free, ANSI
For Use With (Application) Non-pyrogenic, Maxymum Recovery™, used for stringent specifications under strict process controls
For Use With (Equipment) Beckman™ Coulter™ Biomek™ FX, Biomek™ NX, Biomek™ 3000 and Multimek™ automated liquid handling instruments with AP-96, SPAN-8, MP20, MP200 and AP-3

  • Compatible with Beckman Biomek FX and Beckman Multimek instruments with an AP96 head
  • Unsurpassed straightness to ensure tip enters the well smoothly, every time
  • 384/1536-well compatible to meet your most demanding applications

Documentation