Learn More
Axygen™ Biomek™ FX/NX Robotic Tips
Brand:  Axygen FX-250-R-S
Code : A0
Additional Details : Weight : 9.75000kg
 View more versions of this product
Product Code. 12549177
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Product Support team on 01509 555888 for assistance
View alternative products »
Description
Description
  • Compatible with Beckman Biomek FX and Beckman Multimek instruments with an AP96 head
  • Unsurpassed straightness to ensure tip enters the well smoothly, every time
  • 384/1536-well compatible to meet your most demanding applications
Specifications
Specifications
Rack
Beckman Coulter™ Multimek™ SPAN-8
Automation
Sterile
Blue
PP (Polypropylene)
DNase, Rnase Free
Beckman Coulter™ Biomek™ 3000, Beckman Coulter™ Biomek™ FX, Beckman Coulter™ Biomek™ NX, Beckman Coulter™ Multimek™ AP96, Beckman Coulter™ Multimek™ MP20, Beckman Coulter™ Multimek™ MP200, Beckman Coulter™ Multimek™ SPAN-8
96 Tips/Rack
Rack
250μL
Clear
Round
ANSI
Non-pyrogenic, used for stringent specifications under strict process controls
Videos