missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Axygen™ Biomek™ FX/NX Aerosol Barrier Filtered Robotic Tips
Clear polypropylene filter tips; autoclavable.
£96.44 - £7050.00
Specifications
Filtered | Filtered |
---|---|
Color | Clear |
Autoclavable | Not Autoclavable |
Clean Claims | DNase, RNase and Pyrogen free |
For Use With (Application) | Non-pyrogenic, used for stringent specifications under strict process controls |
Product Code | Brand | Volume (Metric) | Sterility | Tip Style | Format | Price | Quantity & Availability | ||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Volume (Metric) | Sterility | Tip Style | Format | Price | Quantity & Availability | ||
13141774
|
Axygen™
FXF-20-R-S |
20 μL | Sterile | Automation, Round | Rack, 96 |
£615.00
Pack of 4800 |
In Stock
Log in to see stock. |
||
15511393
|
Axygen™
FXF-384-30FP-R-S |
30 μL | Sterile | Automation, Fine Point | Rack, 384-well |
£7050.00
Pack of 19200 |
Estimated Shipment: 08-07-2024
Log in to see stock. |
||
12776699
|
Axygen™
FXF-384-XL-R-S |
30 μL | Sterile | Automation, Extended Length | Rack, 384-well |
£5590.00
Pack of 19200 |
Estimated Shipment: 27-08-2024
Log in to see stock. |
||
12796699
|
Axygen™
FXF-50-R-S |
50 μL | Sterile | Automation, Round | Rack, 96 |
£672.00
Pack of 4800 |
Estimated Shipment: 10-09-2024
Log in to see stock. |
||
13187444
|
Axygen™
FXF-180-R-S |
180 μL | Sterile | Automation, Round | Rack, 96 |
£615.00
Pack of 4800 |
Estimated Shipment: 17-05-2024
Log in to see stock. |
||
12786689
|
Axygen™
FXF-180-L-R-S |
180 μL | Sterile | Automation, Round | Rack, 96 |
£685.00
Pack of 4800 |
Estimated Shipment: 01-07-2024
Log in to see stock. |
||
10312440
|
Axygen™
FXF-200-R-S |
200 μL | Sterile | Automation, Round | Rack, 96 |
£631.00
Pack of 4800 |
In Stock
Log in to see stock. |
||
12756699
|
Axygen™
FXF-384-R-S |
25 μL | Sterile | Automation, Round | Rack, 384-well |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||
11346304
|
Axygen™
FXF-384-R-S |
25 μL | Sterile | Automation, Round | Rack, 384-well |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||
11366304
|
Axygen™
FXF-384-XL-R-S |
30 μL | Sterile | Automation, Extended Length | Rack, 384-well |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||
12569187
|
Axygen™
FXF-20-L-R-S |
20 μL | Sterile | Automation, Round | Rack, 96 |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||
11356304
|
Axygen™
FXF-384-XL-L-R-S |
30 μL | Sterile | Automation, Extended Length, Low Retention | Rack, 384-well |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||
12786699
|
Axygen™
FXF-50-L-R-S |
50 μL | Sterile | Automation, Round | Rack, 96 |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||
12589187
|
Axygen™
FXF-50-L-R-S |
50 μL | Sterile | Automation, Round | Rack, 96 |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||
12599187
|
Axygen™
FXF-50-R-S |
50 μL | Sterile | Automation, Round | Rack, 96 |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||
12716699
|
Axygen™
FXF-20-L-R-S |
20 μL | Sterile | Automation, Round | Rack, 96 |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||
11326304
|
Axygen™
FXF-165-WB-R-S |
165 μL | Sterile | Automation, Wide Orifice | Rack, 96 |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||
12549187
|
Axygen™
FXF-180-R-S |
180 μL | Sterile | Automation, Round | Rack, 96 |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||
12579187
|
Axygen™
FXF-20-R-S |
20 μL | Sterile | Automation, Round | Rack, 96 |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||
12746699
|
Axygen™
FXF-384-L-R-S |
30 μL | Sterile | Automation, Round | Rack, 384-well |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||
12539187
|
Axygen™
FXF-180-L-R-S |
180 μL | Sterile | Automation, Round | Rack, 96 |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||
11336304
|
Axygen™
FXF-384-L-R-S |
30 μL | Sterile | Automation, Round | Rack, 384-well |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||
12559187
|
Axygen™
FXF-200-R-S |
200 μL | Sterile | Automation, Round | Rack, 96 |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||
Description
- Tip volume range: 5 to 250 μL
- Tip material: Clear polypropylene
- RNase-/DNase-/human gDNA-free
- non-pyrogenic
- Available options:
- With or without aerosol-resistant filters
- non-sterile or sterile
- Standard or Axygen™ Maxymum Recovery™ surface
- Rack format: ANSI/SLAS
- Rack material: Polypropylene
Specifications
Filtered | |
Not Autoclavable | |
Non-pyrogenic, used for stringent specifications under strict process controls | |
Beckman Biomek |
Clear | |
DNase, RNase and Pyrogen free | |
10 Racks/Pk, 5 Pks/Cs. | |
Polypropylene |