Learn More
Axygen™ Beckman™ Biomek 1000 and 2000 Robotic Tips
Maxymum Recovery™ reduces sample loss and improves reproducibility and accuracy
Brand:  Axygen BTF-50-L-R-S
UNSPSC : 41121608
Code : 68
Additional Details : Weight : 10.40000kg
 View more versions of this product
Product Code. 12782826
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Product Support team on 01509 555888 for assistance
Specifications
Specifications
Filtered Robotic Tip
Sterile
Gray
Round
10 x 96/Rack
RNase, DNase-free, Non-ANSI
Beckman™ Coulter™ Biomek™ 1000 and 2000 series automation systems
50μL
Clear
Filter, Maxymum Recovery
Polypropylene
96 Tips/Rack
Maxymum Recovery™, non-pyrogenic, uses unique diamond polished molding technology, use no mold-releasing agents, nucleic acid, nuclease and endotoxin-safe for complete protection against contamination, minimize waste of precious samples