Learn More
Axygen™ Beckman™ Biomek 1000 and 2000 Robotic Tips
Maxymum Recovery™ reduces sample loss and improves reproducibility and accuracy
Brand:  Axygen BTF-20-R-S
Code : 68
Additional Details : Weight : 10.21000kg
 View more versions of this product
Product Code. 12772826
  • £666.70 / Pack of 4800
Estimated Shipment
06-07-2018
Add to basket
Description
Description
Robotic tips uses unique diamond-polished molding technology use no mold-releasing agents. Tips minimize waste of precious samples and reagents. Nucleic acid, nuclease and endotoxin-safe for complete protection against contamination.
  • Volume: 20 to 1000μL
  • Tip color: Clear
  • Rack color: Grey and Black
  • Available in Maxymum Recovery, Racked, Maxymum Recovery, Racked, Presterilized, Racked and Racked, Presterilized tip types
Specifications
Specifications
10 x 96/Rack
Beckman Coulter™ Biomek™ 1000/2000
Automation
Filtered
Clear
Round
Non-ANSI
Non-pyrogenic, uses unique diamond polished molding technology, use no mold-releasing agents, nucleic acid, nuclease and endotoxin-safe for complete protection against contamination, minimize waste of precious samples and reagents
96 Tips/Rack
Rack
20μL
Sterile
Gray
PP (Polypropylene)
DNase, Rnase Free
Beckman Coulter™ Biomek™ 1000/2000
Videos