Learn More
Axygen™ Beckman™ Biomek 1000 and 2000 Robotic Tips
Maxymum Recovery™ reduces sample loss and improves reproducibility and accuracy
Brand:  Axygen BTF-180-L-R-S
UNSPSC : 41121608
Code : 68
Additional Details : Weight : 10.21000kg
 View more versions of this product
Product Code. 12742826
  • £666.40 / Pack of 4800
Call for latest availability
+44 (0)1509 555 500
Add to basket
Specifications Description & Specifications
Specifications
Filtered Robotic Tip
Sterile
Black
Round
10 x 96/Rack
RNase, DNase-free, Non-ANSI
Beckman™ Coulter™ Biomek™ 1000 and 2000 series automation systems
165μL
Clear
Filter, Maxymum Recovery
Polypropylene
96 Tips/Rack
Maxymum Recovery™, non-pyrogenic, uses unique diamond polished molding technology, use no mold-releasing agents, nucleic acid, nuclease and endotoxin-safe for complete protection against contamination, minimize waste of precious samples