Learn More
Axygen™ Beckman™ Biomek 1000 and 2000 Robotic Tips
Maxymum Recovery™ reduces sample loss and improves reproducibility and accuracy
Brand:  Axygen BTF-50-L-R-S
Code : A0
Additional Details : Weight : 10.40000kg
 View more versions of this product
Product Code. 11366274
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Product Support team on 01509 555888 for assistance
Description
Description
Order Info
Rack: 96 tips/rack, 10 racks/pack, 5 packs/case
Bulk Packed: 1000 tips/unit, 10 Units/case
BT-1000 series:
Rack: 72 tips/rack, 5 racks/case
Specifications
Specifications
Beckman Coulter™ Biomek™ 1000/2000
Automation
Filtered
Clear
Round
Non-ANSI
Maxymum Recovery™, non-pyrogenic, uses unique diamond polished molding technology, use no mold-releasing agents, nucleic acid, nuclease and endotoxin-safe for complete protection against contamination, minimize waste of precious samples
Rack
50μL
Sterile
Gray
PP (Polypropylene)
DNase, Rnase Free
Beckman Coulter™ Biomek™ 1000/2000
Videos