Axygen™ Beckman™ Biomek 1000 and 2000 Robotic Tips

Maxymum Recovery™ reduces sample loss and improves reproducibility and accuracy

Overview
Brand: Axygen

Manufacturer Part Number: BTF-20-L-R-S

X960 20µL MX FILT TIP RACK PST

UNSPSC: 41121608

Code: A0

Additional Details:
Additional Details: Weight: 10.21000kg



Product Code. 11346274

 N/A
N/A

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Product Support team on 01509 555888 for assistance


Description and Specification

Specification

Packaging 10 x 96/Rack
Product Type Filtered Robotic Tip
Material Polypropylene
Quantity 96 Tips/Rack
For Use With (Equipment) Beckman™ Coulter™ Biomek™ 1000 and 2000 series automation systems
Sterility Sterile
Certifications/Compliance RNase, DNase-free, Non-ANSI
Color Clear
Rack Color Gray
Tip Style Filter, Maxymum Recovery
Shape Round
For Use With (Application) Maxymum Recovery™, non-pyrogenic, uses unique diamond polished molding technology, use no mold-releasing agents, nucleic acid, nuclease and endotoxin-safe for complete protection against contamination, minimize waste of precious samples
Volume (Metric) 20μL