missing translation for 'onlineSavingsMsg'
Learn More

Axygen™ Axypet-16 Pipette Tips

For 1536-well pipetting

£860.86 - £5175.00

Specifications

Volume (Metric) 0.5 to 30 μL
Color Clear
Clean Claims DNase, RNase Free
For Use With (Application) For 1536-well pipetting
Format Rack
View More Specs

Products 3
Product Code Brand Sterility Price Quantity & Availability  
Product Code Brand Sterility Price Quantity & Availability  
12792936
View Documents Promotion Details
Axygen™
T-1536-L-R
Non-sterile This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
11344245
View Documents Promotion Details
Axygen™
T-1536-L-R-S
Sterile This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
12702946
View Documents Promotion Details
Axygen™
T-1536-L-R-S
Sterile This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
Description

Description

Axygen™ Universal Tips are designed to fit a wide variety of single and multi-channel pipettors, compatible with most popular brands of pipettors. These polypropylene tips are nonpyrogenic and certified to be RNase-/DNase-free. They are available in Maxymum Recovery, beveled, nonsterile, sterile, non-filtered, and other features for specialized applications. Flexible walls and a series of internal sealing rings ensure a secure fit with less force required to load and eject.

  • These polypropylene tips are nonpyrogenic and certified to be RNase-/DNase-free.
  • Nonpyrogenic
  • Maxymum Recovery technology for ultra-low retention and high yield
  • Sterile
Specifications

Specifications

0.5 to 30 μL
DNase, RNase Free
Rack
1536-Well Pipetting
Clear
For 1536-well pipetting
Low Retention
Polypropylene
Videos
SDS
Documents

Documents

Product Certifications