Learn More
Axygen™ AxyMats™ Sealing Mats for 500uL 96 Well Plates
Minimize evaporation in wide range of temperatures in PCR and storage applications
Brand:  Axygen™ AM-500UL-RD-S
Code : A0
Additional Details : Weight : 1.40000kg
 View more versions of this product
Product Code. 12551328
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Description
Description
Axygen AxyMat sealing mats can be used in a wide range of temperatures for PCR and storage applications. AxyMats minimize evaporation and are made of research-grade silicone which can be autoclaved. They can be reused following a bleach wash and ethanol rinse protocol.
Specifications
Specifications
Natural
Sterile
96
10
Sealing Mat
Silicone
Autoclavable
5
No
96-well Deep Well Plates with V-bottom Wells, 500 μL