Axygen™ 96-Well PCR Microplates

Available in a variety of colors

Overview
Brand: Axygen

Manufacturer Part Number: PCR-96-A

X50 PCR microplate Axygen 96 well polypropylene

UNSPSC: 41122102

Code: 68

Additional Details:
Additional Details: Weight: 0.90000kg



Product Code. 12789438

 N/A
N/A

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Product Support team on 01509 555888 for assistance


  • Description and Specification
  • Alternative Products
Description and Specification

Specification

Well Color Clear
Material Polypropylene
Sterile No
Product Type 96-Well PCR Microplate
Volume (Metric) Well 200μL
Color Assorted
Material Polypropylene
Applications PCR thermal transfer, RT-PCR applications
For Use With (Application) PCR thermal transfer, RT-PCR applications
Certifications/Compliance RNase, DNase-free
Sterile No
Alternative Products