Learn More
Corning™ Axygen™ 96-well Low Profile PCR Microplates
Constructed with ultra-thin and consistent wall thickness to allow precise thermal transfer for optimum results. Corning™ Axygen™ 96-well PCR Microplates are the ideal option for use in PCR thermal transfer and Real-Time PCR applications. The low profile design results in faster and more efficient PCR reactions.
Brand: Corning™ PCR-96-LP-FLT-W
Code : A0
Additional Details : Weight : 1.95000kg
Description
- Low profile; Optimized for fast PCR
- Certified RNase-/DNase-free
- Nonpyrogenic, nonsterile
- Sealing options: Axymats, heat sealing, adhesive films and foils, flat and domed cap strips
Specifications
Flat top | |
RNase-/DNase-free | |
PCR thermal transfer, RT-PCR applications | |
Polypropylene | |
Non-sterile | |
Round |
No | |
White | |
0.1mL thermal cycler blocks; Standard Biometra thermal cyclers, Eppendorf MasterCycler qPCR, MJ Opticon/Chromo4 qPCR | |
Non-skirted | |
100μL |