Axygen™ 384-well Skirted PCR Microplates

Single and double notch 384-well PCR plates

Overview
Brand: Axygen

Manufacturer Part Number: PCR-384-LC480-W

X10 384 WELL PCR MICROPLATE

UNSPSC: 41122102

Code: A0

Additional Details:
Additional Details: Weight: 1.40000kg



Product Code. 12596877

Quantity Price
1 £ 79.23 / Pack of 10
Call for latest availability
+44 (0)1509 555 500
Add to basket


Description and Specification

Specification

Volume (Metric) Well 55μL
For Use With (Application) PCR
For Use With (Equipment) For Roche light cycler 480
Includes Sealing film, single notch
Well Color White
Material Polypropylene
Sterile No
Certifications/Compliance RNase, DNase-free
Color White
Volume (Metric) Working 30μL
Well Design Round
Material Polypropylene
Skirt Style Full
Sterile No
Well Shape Round
Applications PCR
Product Type 384-Well PCR Microplate