Learn More
Axygen™ Maxymum Recovery™ 300 μL Universal Pipetter Tips
Maxymum Recovery tips fit popular brands of pipettes
Brand:  Axygen™ T-350-C-L-R
Code : 68
Additional Details : Weight : 8.16000kg
 View more versions of this product
Product Code. 12716609
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Description
Description
Axygen™ Universal Tips are designed to fit a wide variety of single and multi-channel pipettors, compatible with most popular brands of pipettors. These polypropylene tips are nonpyrogenic and certified to be RNase-/DNase-free. They are available in Maxymum Recovery, beveled, nonsterile, sterile, non-filtered, and other features for specialized applications. Flexible walls and a series of internal sealing rings ensure a secure fit with less force required to load and eject.
  • These polypropylene tips are nonpyrogenic and certified to be RNase-/DNase-free.
  • Nonpyrogenic
  • Fine-Point
  • Maxymum Recovery technology for ultra-low retention and high yield
Order Info
Bulk: 1000 tips/pack, 240 packs/case
Racked: 96 tips/rack, 10 racks/pack, 5 packs/case
Specifications
Specifications
1 to 300 μL
Clear
Rack
Hinged rack
Maximum Recovery
Non-sterile
DNase, RNase, ATP, and Endotoxin Free
Low Retention, Universal
Universal Fit
Polypropylene