Learn More
Axygen™ 250μL Automation Tips for Agilent™/Velocity 11 VPrep™ and Bravo, 96 Tip Format
For use with Agilent/Velocity11 automation systems
Brand:  Axygen VT-250-R-LAB-S
Code : 68
Additional Details : Weight : 10.00000kg
 View more versions of this product
Product Code. 12726659
  • £326.76 / Pack of 4800
Estimated Shipment
06-07-2018
Add to basket
Specifications
Specifications
Rack
Agilent™/Velocity VPrep® 8LT├żAgilent™/Velocity VPrep® 96LT├żAgilent™ Bravo® 96LT
Automation
Sterile
Round
ANSI
Non-pyrogenic, used for stringent specifications under strict process controls
96 Tips/Rack
Rack
250μL
Clear
PP (Polypropylene)
DNase, Rnase Free
Agilent™/Velocity VPrep® 8LT, Agilent™/Velocity VPrep® 96LT, Agilent™ Bravo® 96LT
Videos