missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Axygen™ 200μL Universal Pipette Tips, Beveled, Reference, Maxymum Recovery
Designed to fit a wide variety of single and multi-channel pipettes
Brand: Axygen™ TR-222-C-L-STK-S
Code : 68
Additional Details : Weight : 4.10000kg
Description
- Optimized tip bevel reduces sample hang-up, improves pipetting accuracy
- Maxymum recovery allows complete retrieval of precious samples, saves costly reagents
- Wide bore tips of orifice size: 0.062 in. (1.52mm) makes it ideal for pipetting viscous materials, genomic DNA or delicate cell cultures
- Graduation marks allow a visual confirmation of pipettor draw volume
- Presterilized tips are e-beam irradiated to prevent product degradation
- Clean-room manufacturing environment certifies products safe from RNase, DNase and pyrogen safe
Specifications
1 to 200 μL | |
Clear | |
PP (Polypropylene) | |
Universal Fit Pipettor | |
Rack |
Sterile | |
Autoclavable | |
DNase, RNase, and Pyrogen free | |
Universal Fit Pipettor | |
Beveled, Low Retention, Universal |