missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Axygen™ 200μL Universal Pipette Tips, Beveled, Reference, Maxymum Recovery
Designed to fit a wide variety of single and multi-channel pipettes
Brand: Axygen™ TR-222-C-L-R
Code : 68
Additional Details : Weight : 8.16000kg
Description
- Optimized tip bevel reduces sample hang-up, improves pipetting accuracy
- Maxymum recovery allows complete retrieval of precious samples, saves costly reagents
- Wide bore tips of orifice size: 0.062 in. (1.52mm) makes it ideal for pipetting viscous materials, genomic DNA or delicate cell cultures
- Graduation marks allow a visual confirmation of pipettor draw volume
- Presterilized tips are e-beam irradiated to prevent product degradation
- Clean-room manufacturing environment certifies products safe from RNase, DNase and pyrogen safe
Specifications
1 to 200 μL | |
Clear | |
DNase, RNase, DNA, and PCR inhibitor Free | |
Beveled, Low Retention, Universal | |
Yes | |
10, 50, 100μL |
Non-sterile | |
Autoclavable | |
Rack | |
Universal Fit | |
Polypropylene |