missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Axygen™ 200μL Universal Pipette Tips: 200μL, Beveled
Designed to fit a wide variety of single and multi-channel pipettes.
Brand: Axygen™ T-200-C-R-S
Code : NEW
Additional Details : Weight : 8.05000kg
Description
- Optimized tip bevel reduces sample hang-up, improves pipetting accuracy
- Wide bore tips of orifice size: 0.062 in. (1.52 mm) makes it ideal for pipetting viscous materials, genomic DNA or delicate cell cultures
- Graduation marks allow a visual confirmation of pipettor draw volume
- Presterilized tips are e-beam irradiated to prevent product degradation
- Clean-room manufacturing environment certifies products safe from RNase, DNase and pyrogen safe
- Also available with Maxymum recovery, which allows complete retrieval of precious samples, saves costly reagents
Specifications
1 to 200 μL | |
Clear | |
DNase, RNase, and Pyrogen free | |
Beveled, Universal | |
Yes |
Sterile | |
Autoclavable | |
Hinged Rack | |
Universal Fit | |
Polypropylene |