missing translation for 'onlineSavingsMsg'
Learn More

Axygen™ 200μL Universal Pipetter Tips: 200μL, Beveled, Maxymum Recovery

Designed to fit a wide variety of single and multi-channel pipettes

Brand:  Axygen T-200-C-L-R

Code : 68

Additional Details : Weight : 8.05000kg

 View more versions of this product

Product Code. 10506120

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Product Support team on 01509 555888 for assistance
View alternative products »


Description

Description

  • Optimized tip bevel reduces sample hang-up, improves pipetting accuracy
  • Maxymum recovery allows complete retrieval of precious samples, saves costly reagents
  • Wide bore tips of orifice size: 0.062 in. (1.52mm) makes it ideal for pipetting viscous materials, genomic DNA or delicate cell cultures
  • Graduation marks allow a visual confirmation of pipettor draw volume
  • Presterilized tips are e-beam irradiated to prevent product degradation
  • Clean-room manufacturing environment certifies products safe from RNase, DNase and pyrogen safe

Specifications

Specifications

Universal Pipetter Tips
Nonsterile
Yes
Polypropylene
DNA, DNase, RNase and PCR inhibitor-free
1 to 200μL
Clear
Beveled; Maxymum Recovery
Rack
Wide variety of single and multichannel pipetters
Videos